3-Chloro-4-ethoxy-2-methoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

61676-500MG 61676-1G 61676-5G
77339-662EA 611.82 USD
77339-662 77355-602 77379-650
3-Chloro-4-ethoxy-2-methoxybenzoic acid ≥97%
3-Chloro-4-ethoxy-2-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₁ClO₄
Storage Temperature: Ambient
MDL Number: MFCD34760482
CAS Number: 2386780-97-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR