2-(Benzyloxy)-1-bromo-3-chloro-4-ethoxybenzene ≥97%

Supplier: AOBChem USA

Warning

61673-500MG 61673-1G 61673-5G 61673-25G
77331-334EA 281.19 USD
77331-334 77355-598 77367-476 77388-978
2-(Benzyloxy)-1-bromo-3-chloro-4-ethoxybenzene ≥97%
2-(Benzyloxy)-1-bromo-3-chloro-4-ethoxybenzene

Precaución: for Research Use Only

Formula: C₁₅H₁₄BrClO₂
MW: 341.6274 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760479
CAS Number: 2764731-26-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR