1-Bromo-3-chloro-2,4-diethoxybenzene ≥97%

Supplier: AOBChem USA

Warning

61672-500MG 61672-1G 61672-5G 61672-25G
77331-332EA 279.13 USD
77331-332 77355-596 77367-474 77388-976
1-Bromo-3-chloro-2,4-diethoxybenzene ≥97%
1-Bromo-3-chloro-2,4-diethoxybenzene

Precaución: for Research Use Only

Formula: C₁₀H₁₂BrClO₂
MW: 279.5580 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760478
CAS Number: 685143-15-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR