(2,5-Dichloro-4-fluorophenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

61667-500MG 61667-1G 61667-5G
77335-444EA 353.29 USD
77335-444 77355-592 77379-644
(2,5-Dichloro-4-fluorophenyl)(methyl)sulfane ≥97%
(2,5-Dichloro-4-fluorophenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₇H₅Cl₂FS
MW: 211.0840 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28786341
CAS Number: 1803777-45-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR