1,4-Dichloro-2-fluoro-5-iodobenzene ≥97%

Supplier: AOBChem USA

Warning

61666-500MG 61666-1G 61666-5G
77331-328EA 319.3 USD
77331-328 77355-590 77379-642
1,4-Dichloro-2-fluoro-5-iodobenzene ≥97%
1,4-Dichloro-2-fluoro-5-iodobenzene

Precaución: for Research Use Only

Formula: C₆H₂Cl₂FI
MW: 290.8889 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28786117
CAS Number: 1393843-49-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR