(2-Bromo-3-ethoxyphenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

61658-500MG 61658-1G 61658-5G 61658-25G
77331-324EA 157.59 USD
77331-324 77345-650 77367-466 77388-970
(2-Bromo-3-ethoxyphenyl)methanol ≥97%
(2-Bromo-3-ethoxyphenyl)methanol

Precaución: for Research Use Only

Formula: C₉H₁₁BrO₂
MW: 231.0864 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760473
CAS Number: 1934739-79-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR