Ethyl 5-isopropyl-2-methoxybenzoate ≥97%

Supplier: AOBChem USA

Warning

61654-500MG 61654-1G 61654-5G
77331-320EA 342.99 USD
77331-320 77355-584 77379-638
Ethyl 5-isopropyl-2-methoxybenzoate ≥97%
Ethyl 5-isopropyl-2-methoxybenzoate

Precaución: for Research Use Only

Formula: C₁₃H₁₈O₃
MW: 222.2802 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760469
CAS Number: 1892396-94-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR