Methyl 5-isopropyl-2-methoxybenzoate ≥97%

Supplier: AOBChem USA

Warning

61653-500MG 61653-1G 61653-25G 61653-5G
77331-318EA 284.28 USD
77331-318 77355-582 77388-966 77379-636
Methyl 5-isopropyl-2-methoxybenzoate ≥97%
Methyl 5-isopropyl-2-methoxybenzoate

Precaución: for Research Use Only

Formula: C₁₂H₁₆O₃
Storage Temperature: Ambient
MDL Number: MFCD17167549
CAS Number: 87188-68-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR