(5-Iodo-2-(methylthio)phenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

61652-500MG 61652-1G 61652-5G 61652-25G
77331-316EA 126.69 USD
77331-316 77344-176 77367-464 77379-634
(5-Iodo-2-(methylthio)phenyl)methanol ≥97%
(5-Iodo-2-(methylthio)phenyl)methanol

Precaución: for Research Use Only

Formula: C₈H₉IOS
MW: 280.1259 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760468
CAS Number: 2764732-36-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR