3-Bromo-2,6-dimethoxy-5-(p-tolyl)pyridine ≥97%

Supplier: AOBChem USA

Warning

61648-500MG 61648-1G 61648-5G 61648-25G
77331-314EA 208.06 USD
77331-314 77355-580 77367-462 77388-964
3-Bromo-2,6-dimethoxy-5-(p-tolyl)pyridine ≥97%
3-Bromo-2,6-dimethoxy-5-(p-tolyl)pyridine

Precaución: for Research Use Only

Formula: C₁₄H₁₄BrNO₂
Storage Temperature: Ambient
MDL Number: MFCD34760464
CAS Number: 2764728-79-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR