(2-Chloro-6-iodo-3-methylphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

61646-500MG 61646-1G 61646-5G 61646-25G
77331-310EA 125.66 USD
77331-310 77344-094 77367-458 77379-632
(2-Chloro-6-iodo-3-methylphenyl)(methyl)sulfane ≥97%
(2-Chloro-6-iodo-3-methylphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₈H₈ClIS
MW: 298.5715 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760462
CAS Number: 2711231-76-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR