3-Chloro-4-methyl-2-(methylthio)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

61643-500MG 61643-1G 61643-5G 61643-25G
77331-308EA 140.08 USD
77331-308 77344-724 77367-456 77388-960
3-Chloro-4-methyl-2-(methylthio)benzoic acid ≥97%
3-Chloro-4-methyl-2-(methylthio)benzoic acid

Precaución: for Research Use Only

Formula: C₉H₉ClO₂S
Storage Temperature: Ambient
MDL Number: MFCD34760459
CAS Number: 2764729-27-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR