Ethyl 3,5-dimethyl-4-(methylthio)benzoate ≥95%

Supplier: AOBChem USA

Warning

61642-500MG 61642-1G 61642-5G
77335-442EA 353.29 USD
77335-442 77355-576 77379-630
Ethyl 3,5-dimethyl-4-(methylthio)benzoate ≥95%
Ethyl 3,5-dimethyl-4-(methylthio)benzoate

Precaución: for Research Use Only

Formula: C₁₂H₁₆O₂S
Storage Temperature: Ambient
MDL Number: MFCD34760458
CAS Number: 2764731-83-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR