3-Chloro-2-fluoro-6-formylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

61631-500MG 61631-1G 61631-5G 61631-25G
77331-304EA 204.97 USD
77331-304 77355-572 77367-452 77388-956
3-Chloro-2-fluoro-6-formylbenzoic acid ≥97%
3-Chloro-2-fluoro-6-formylbenzoic acid

Precaución: for Research Use Only

Formula: C₈H₄ClFO₃
MW: 202.5669 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760449
CAS Number: 2384182-29-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR