1-(2,6-Difluoro-3-(methylthio)phenyl)-2,2,2-trifluoroethanone ≥97%

Supplier: AOBChem USA

Warning

61619-500MG 61619-1G 61619-25G 61619-5G
77331-302EA 287.37 USD
77331-302 77355-568 77388-954 77379-624
1-(2,6-Difluoro-3-(methylthio)phenyl)-2,2,2-trifluoroethanone ≥97%
1-(2,6-Difluoro-3-(methylthio)phenyl)-2,2,2-trifluoroethanone

Precaución: for Research Use Only

Formula: C₉H₅F₅OS
Storage Temperature: Ambient
MDL Number: MFCD34760443
CAS Number: 2755722-58-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR