(3-Bromo-2,4-difluorophenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

61617-500MG 61617-1G 61617-5G 61617-25G
77331-300EA 166.86 USD
77331-300 77346-220 77367-450 77388-952
(3-Bromo-2,4-difluorophenyl)(methyl)sulfane ≥97%
(3-Bromo-2,4-difluorophenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₇H₅BrF₂S
MW: 239.0804 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29089635
CAS Number: 2092059-70-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR