(4-Chloro-2-fluoro-3-methoxy-5-methylphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

61613-500MG 61613-1G 61613-5G 61613-25G
77331-292EA 136.99 USD
77331-292 77344-550 77367-442 77388-946
(4-Chloro-2-fluoro-3-methoxy-5-methylphenyl)(methyl)sulfane ≥97%
(4-Chloro-2-fluoro-3-methoxy-5-methylphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₉H₁₀ClFOS
Storage Temperature: Ambient
MDL Number: MFCD34760439
CAS Number: 2755716-91-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR