4-Chloro-2-fluoro-3-methoxy-5-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

61608-500MG 61608-1G 61608-5G 61608-25G
77331-288EA 82.4 USD
77331-288 77342-158 77367-438 77379-620
4-Chloro-2-fluoro-3-methoxy-5-methylbenzoic acid ≥97%
4-Chloro-2-fluoro-3-methoxy-5-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₉H₈ClFO₃
MW: 218.6094 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760434
CAS Number: 1895485-76-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR