3-(5-Bromo-2-fluorophenyl)-3-hydroxypropanoic acid ≥95%

Supplier: AOBChem USA

Warning

61577-500MG 61577-1G 61577-5G
77339-614EA 603.58 USD
77339-614 77355-564 77379-618
3-(5-Bromo-2-fluorophenyl)-3-hydroxypropanoic acid ≥95%
3-(5-Bromo-2-fluorophenyl)-3-hydroxypropanoic acid

Precaución: for Research Use Only

Formula: C₉H₈BrFO₃
MW: 263.0604 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16749135
CAS Number: 1249004-30-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR