3-Hydroxy-3-(4-(trifluoromethoxy)phenyl)propanoic acid ≥97%
Supplier: AOBChem USA
|
Warning
|
61571-500MG
61571-1G
61571-5G
61571-25G
77331-284EA
243.08
USD
77331-284
77355-562
77367-434
77388-940
3-Hydroxy-3-(4-(trifluoromethoxy)phenyl)propanoic acid ≥97%
3-Hydroxy-3-(4-(trifluoromethoxy)phenyl)propanoic acid
Precaución: for Research Use Only
Formula:
C₁₀H₉F₃O₄ MW: 250.1712 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD17969602 CAS Number: 1282918-67-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...