3-(4-Bromophenyl)-3-hydroxypropanoic acid ≥97%

Supplier: AOBChem USA

Warning

61569-500MG 61569-1G 61569-5G
77336-700EA 391.4 USD
77336-700 77355-558 77379-614
3-(4-Bromophenyl)-3-hydroxypropanoic acid ≥97%
3-(4-Bromophenyl)-3-hydroxypropanoic acid

Precaución: for Research Use Only

Formula: C₉H₉BrO₃
MW: 245.0699 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16749140
CAS Number: 40620-68-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR