2-Methoxy-2'-methyl-[1,1'-biphenyl]-4-carboxylic acid ≥97%
Supplier: AOBChem USA
|
Warning
|
61540-500MG
61540-1G
61540-5G
61540-25G
77331-282EA
230.72
USD
77331-282
77355-554
77367-432
77388-938
2-Methoxy-2'-methyl-[1,1'-biphenyl]-4-carboxylic acid ≥97%
2-Methoxy-2'-methyl-[1,1'-biphenyl]-4-carboxylic acid
Precaución: for Research Use Only
Formula:
C₁₅H₁₄O₃ MW: 242.2698 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18318916 CAS Number: 175153-24-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...