2-Methoxy-4'-methyl-[1,1'-biphenyl]-4-carboxylic acid ≥97%

Supplier: AOBChem USA

Warning

61530-500MG 61530-1G 61530-5G 61530-25G
77331-278EA 233.81 USD
77331-278 77355-548 77367-430 77388-936
2-Methoxy-4'-methyl-[1,1'-biphenyl]-4-carboxylic acid ≥97%
2-Methoxy-4'-methyl-[1,1'-biphenyl]-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₅H₁₄O₃
MW: 242.2698 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18318961
CAS Number: 1262004-95-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR