Ethyl 6-methyl-[1,1'-biphenyl]-3-carboxylate ≥97%

Supplier: AOBChem USA

Warning

61528-500MG 61528-1G 61528-5G
77338-100EA 464.53 USD
77338-100 77355-546 77379-606
Ethyl 6-methyl-[1,1'-biphenyl]-3-carboxylate ≥97%
Ethyl 6-methyl-[1,1'-biphenyl]-3-carboxylate

Precaución: for Research Use Only

Formula: C₁₆H₁₆O₂
MW: 240.2970 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760389
CAS Number: 2901083-11-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR