N-Cyclopropyl-2-fluoro-5-(trifluoromethyl)benzamide ≥97%

Supplier: AOBChem USA

Warning

61524-500MG 61524-1G 61524-5G 61524-25G
77331-276EA 234.84 USD
77331-276 77355-544 77367-428 77388-934
N-Cyclopropyl-2-fluoro-5-(trifluoromethyl)benzamide ≥97%
N-Cyclopropyl-2-fluoro-5-(trifluoromethyl)benzamide

Precaución: for Research Use Only

Formula: C₁₁H₉F₄NO
Storage Temperature: Ambient
MDL Number: MFCD24793064
CAS Number: 1516702-40-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR