N-Cyclohexyl-2-fluoro-5-(trifluoromethyl)benzamide ≥97%

Supplier: AOBChem USA

Warning

61523-500MG 61523-1G 61523-5G 61523-25G
77331-274EA 234.84 USD
77331-274 77355-542 77367-426 77388-932
N-Cyclohexyl-2-fluoro-5-(trifluoromethyl)benzamide ≥97%
N-Cyclohexyl-2-fluoro-5-(trifluoromethyl)benzamide

Precaución: for Research Use Only

Formula: C₁₄H₁₅F₄NO
Storage Temperature: Ambient
MDL Number: MFCD34760385
CAS Number: 1506969-61-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR