N,N-diethyl-2-fluoro-5-(trifluoromethyl)benzamide ≥97%

Supplier: AOBChem USA

Warning

61521-1G 61521-5G 61521-25G
77340-778EA 60.77 USD
77340-778 77367-424 77379-604
N,N-diethyl-2-fluoro-5-(trifluoromethyl)benzamide ≥97%
N,N-diethyl-2-fluoro-5-(trifluoromethyl)benzamide

Precaución: for Research Use Only

Formula: C₁₂H₁₃F₄NO
Storage Temperature: Ambient
MDL Number: MFCD24793065
CAS Number: 1528932-52-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR