(2-Fluoro-5-(trifluoromethyl)phenyl)(piperidin-1-yl)methanone ≥97%

Supplier: AOBChem USA

Warning

61520-500MG 61520-1G 61520-5G 61520-25G
77331-272EA 230.72 USD
77331-272 77355-540 77367-422 77388-930
(2-Fluoro-5-(trifluoromethyl)phenyl)(piperidin-1-yl)methanone ≥97%
(2-Fluoro-5-(trifluoromethyl)phenyl)(piperidin-1-yl)methanone

Precaución: for Research Use Only

Formula: C₁₃H₁₃F₄NO
MW: 275.2420 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23374253
CAS Number: 1542461-01-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR