tert-Butyl 2,6-dichloro-4-(trifluoromethoxy)benzoate ≥97%

Supplier: AOBChem USA

Warning

61508-500MG 61508-1G 61508-5G 61508-25G
77331-270EA 172.01 USD
77331-270 77346-424 77367-410 77388-928
tert-Butyl 2,6-dichloro-4-(trifluoromethoxy)benzoate ≥97%
tert-Butyl 2,6-dichloro-4-(trifluoromethoxy)benzoate

Precaución: for Research Use Only

Formula: C₁₂H₁₁Cl₂F₃O₃
MW: 331.1151 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760382
CAS Number: 2807440-78-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR