2-(Benzyloxy)-3,5-dichlorobenzoic acid ≥97%

Supplier: AOBChem USA

Warning

61501-500MG 61501-1G 61501-5G 61501-25G
77331-266EA 249.26 USD
77331-266 77355-536 77367-406 77388-924
2-(Benzyloxy)-3,5-dichlorobenzoic acid ≥97%
2-(Benzyloxy)-3,5-dichlorobenzoic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₀Cl₂O₃
MW: 297.1334 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04542750
CAS Number: 518978-12-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR