3,5-Dichloro-2-isopropoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

61494-500MG 61494-1G 61494-5G
77337-466EA 414.06 USD
77337-466 77355-534 77379-592
3,5-Dichloro-2-isopropoxybenzoic acid ≥97%
3,5-Dichloro-2-isopropoxybenzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₀Cl₂O₃
MW: 249.0906 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12189243
CAS Number: 1154658-67-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR