6-(Benzyloxy)-3-fluoro-2-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

61481-500MG 61481-1G 61481-5G
77338-934EA 532.51 USD
77338-934 77355-530 77379-588
6-(Benzyloxy)-3-fluoro-2-methylbenzoic acid ≥97%
6-(Benzyloxy)-3-fluoro-2-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₁₅H₁₃FO₃
MW: 260.2603 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760368
CAS Number: 2807467-48-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR