(2-Chloro-4-methoxy-3-methylphenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

61480-500MG 61480-1G 61480-25G 61480-5G
77331-264EA 289.43 USD
77331-264 77355-528 77388-922 77379-586
(2-Chloro-4-methoxy-3-methylphenyl)methanol ≥97%
(2-Chloro-4-methoxy-3-methylphenyl)methanol

Precaución: for Research Use Only

Formula: C₉H₁₁ClO₂
MW: 186.6354 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760367
CAS Number: 2385394-08-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR