6-(Benzyloxy)-3-fluoro-2-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

61475-500MG 61475-1G 61475-5G
77338-616EA 503.67 USD
77338-616 77355-526 77379-584
6-(Benzyloxy)-3-fluoro-2-methylbenzaldehyde ≥97%
6-(Benzyloxy)-3-fluoro-2-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₁₅H₁₃FO₂
Storage Temperature: Ambient
MDL Number: MFCD34760363
CAS Number: 2807450-60-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR