Ethyl 4-bromo-2-methoxy-5-(trifluoromethyl)benzoate ≥97%

Supplier: AOBChem USA

Warning

61468-500MG 61468-1G 61468-5G
77337-188EA 408.91 USD
77337-188 77355-524 77379-582
Ethyl 4-bromo-2-methoxy-5-(trifluoromethyl)benzoate ≥97%
Ethyl 4-bromo-2-methoxy-5-(trifluoromethyl)benzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₀BrF₃O₃
Storage Temperature: Ambient
MDL Number: MFCD34760360
CAS Number: 2705276-79-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR