Methyl 4-bromo-2-methoxy-5-(trifluoromethyl)benzoate ≥97%

Supplier: AOBChem USA

Warning

61467-500MG 61467-1G 61467-5G
77337-186EA 408.91 USD
77337-186 77355-522 77379-580
Methyl 4-bromo-2-methoxy-5-(trifluoromethyl)benzoate ≥97%
Methyl 4-bromo-2-methoxy-5-(trifluoromethyl)benzoate

Precaución: for Research Use Only

Formula: C₁₀H₈BrF₃O₃
MW: 313.0679 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760359
CAS Number: 2415000-75-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR