4-Bromo-2-methoxy-5-(trifluoromethyl)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

61466-500MG 61466-1G 61466-5G
77335-578EA 357.41 USD
77335-578 77355-520 77379-578
4-Bromo-2-methoxy-5-(trifluoromethyl)benzoic acid ≥97%
4-Bromo-2-methoxy-5-(trifluoromethyl)benzoic acid

Precaución: for Research Use Only

Formula: C₉H₆BrF₃O₃
MW: 299.0413 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760358
CAS Number: 2384476-15-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR