4-Bromo-2-methoxy-5-(trifluoromethyl)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

61460-500MG 61460-1G 61460-5G
77336-434EA 387.28 USD
77336-434 77355-518 77379-576
4-Bromo-2-methoxy-5-(trifluoromethyl)benzaldehyde ≥97%
4-Bromo-2-methoxy-5-(trifluoromethyl)benzaldehyde

Precaución: for Research Use Only

Formula: C₉H₆BrF₃O₂
Storage Temperature: Ambient
MDL Number: MFCD34471240
CAS Number: 2351862-91-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR