1-Chloro-2-ethoxy-3-fluoro-4-iodobenzene ≥97%

Supplier: AOBChem USA

Warning

61457-500MG 61457-1G 61457-5G
77331-260EA 322.39 USD
77331-260 77355-514 77379-572
1-Chloro-2-ethoxy-3-fluoro-4-iodobenzene ≥97%
1-Chloro-2-ethoxy-3-fluoro-4-iodobenzene

Precaución: for Research Use Only

Formula: C₈H₇ClFIO
MW: 300.4964 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760352
CAS Number: 2385204-76-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR