1,5-Dichloro-3-iodo-2-isopropoxybenzene ≥97%

Supplier: AOBChem USA

Warning

61453-500MG 61453-1G 61453-5G 61453-25G
77331-252EA 161.71 USD
77331-252 77345-904 77367-398 77388-914
1,5-Dichloro-3-iodo-2-isopropoxybenzene ≥97%
1,5-Dichloro-3-iodo-2-isopropoxybenzene

Precaución: for Research Use Only

Formula: C₉H₉Cl₂IO
MW: 330.9776 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760349
CAS Number: 2568197-66-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR