(3-Iodo-2-methylphenyl)(4-methylpiperazin-1-yl)methanone ≥95%

Supplier: AOBChem USA

Warning

61448-500MG 61448-1G 61448-5G 61448-25G
77331-248EA 109.18 USD
77331-248 77343-240 77367-394 77379-568
(3-Iodo-2-methylphenyl)(4-methylpiperazin-1-yl)methanone ≥95%
(3-Iodo-2-methylphenyl)(4-methylpiperazin-1-yl)methanone

Precaución: for Research Use Only

Formula: C₁₃H₁₇IN₂O
MW: 344.1913 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760346
CAS Number: 2901099-90-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR