(3-Iodo-2-methylphenyl)(morpholino)methanone ≥95%

Supplier: AOBChem USA

Warning

61446-500MG 61446-1G 61446-5G
77331-244EA 332.69 USD
77331-244 77355-508 77379-566
(3-Iodo-2-methylphenyl)(morpholino)methanone ≥95%
(3-Iodo-2-methylphenyl)(morpholino)methanone

Precaución: for Research Use Only

Formula: C₁₂H₁₄INO₂
MW: 331.1495 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32666922
CAS Number: 1269520-72-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR