(3-Iodo-2-methylphenyl)(piperidin-1-yl)methanone ≥97%

Supplier: AOBChem USA

Warning

61445-500MG 61445-1G 61445-5G
77331-242EA 332.69 USD
77331-242 77355-506 77379-564
(3-Iodo-2-methylphenyl)(piperidin-1-yl)methanone ≥97%
(3-Iodo-2-methylphenyl)(piperidin-1-yl)methanone

Precaución: for Research Use Only

Formula: C₁₃H₁₆INO
MW: 329.1767 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32666921
CAS Number: 2749754-93-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR