N-Ethyl-3-iodo-2-methylbenzamide ≥97%

Supplier: AOBChem USA

Warning

61444-500MG 61444-1G 61444-5G
77331-240EA 332.69 USD
77331-240 77355-504 77379-562
N-Ethyl-3-iodo-2-methylbenzamide ≥97%
N-Ethyl-3-iodo-2-methylbenzamide

Precaución: for Research Use Only

Formula: C₁₀H₁₂INO
Storage Temperature: Ambient
CAS Number: 1863037-73-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR