N,N-diethyl-3-iodo-2-methylbenzamide ≥97%

Supplier: AOBChem USA

Warning

61443-500MG 61443-1G 61443-5G
77331-238EA 332.69 USD
77331-238 77355-502 77379-560
N,N-diethyl-3-iodo-2-methylbenzamide ≥97%
N,N-diethyl-3-iodo-2-methylbenzamide

Precaución: for Research Use Only

Formula: C₁₂H₁₆INO
Storage Temperature: Ambient
CAS Number: 1676075-44-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR