(3-Iodo-2-methylphenyl)(pyrrolidin-1-yl)methanone ≥97%

Supplier: AOBChem USA

Warning

61442-500MG 61442-1G 61442-5G
77331-236EA 332.69 USD
77331-236 77355-500 77379-558
(3-Iodo-2-methylphenyl)(pyrrolidin-1-yl)methanone ≥97%
(3-Iodo-2-methylphenyl)(pyrrolidin-1-yl)methanone

Precaución: for Research Use Only

Formula: C₁₂H₁₄INO
MW: 315.1501 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32666919
CAS Number: 1269520-79-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR