N-Cyclopropyl-3-iodo-2-methylbenzamide ≥97%

Supplier: AOBChem USA

Warning

61440-500MG 61440-1G 61440-5G
77331-234EA 332.69 USD
77331-234 77355-498 77379-556
N-Cyclopropyl-3-iodo-2-methylbenzamide ≥97%
N-Cyclopropyl-3-iodo-2-methylbenzamide

Precaución: for Research Use Only

Formula: C₁₁H₁₂INO
Storage Temperature: Ambient
MDL Number: MFCD32666915
CAS Number: 2749754-84-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR