3-Iodo-N,2-dimethylbenzamide ≥97%
Supplier: AOBChem USA
|
Warning
|
61439-500MG
61439-1G
61439-5G
77331-232EA
332.69
USD
77331-232
77355-496
77379-554
3-Iodo-N,2-dimethylbenzamide ≥97%
3-Iodo-N,2-dimethylbenzamide
Precaución: for Research Use Only
Formula:
C₉H₁₀INO Storage Temperature: Ambient |
MDL Number:
MFCD30299379 CAS Number: 1862379-11-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...