3-Iodo-2-methylbenzamide ≥95%

Supplier: AOBChem USA

Warning

61437-500MG 61437-1G 61437-5G
77331-230EA 332.69 USD
77331-230 77355-494 77379-552
3-Iodo-2-methylbenzamide ≥95%
3-Iodo-2-methylbenzamide

Precaución: for Research Use Only

Formula: C₈H₈INO
Storage Temperature: Ambient
MDL Number: MFCD01940320
CAS Number: 52107-88-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR