Methyl 3-ethoxy-5-isopropylbenzoate ≥97%

Supplier: AOBChem USA

Warning

61426-500MG 61426-1G 61426-5G
77337-308EA 410.97 USD
77337-308 77355-492 77379-550
Methyl 3-ethoxy-5-isopropylbenzoate ≥97%
Methyl 3-ethoxy-5-isopropylbenzoate

Precaución: for Research Use Only

Formula: C₁₃H₁₈O₃
MW: 222.2802 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760341
CAS Number: 2764732-12-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR